Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-tert-Butoxy-6-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H55085-06 H55085-14 H55085-03
AAH55085-06EA 82.89 USD InStock
AAH55085-06 AAH55085-14 AAH55085-03
2-tert-Butoxy-6-chloropyridine 97%
2-tert-Butoxy-6-chloropyridine


Formula: C₉H₁₂ClNO
Density: 1.076
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD08705263
CAS Number: 547740-43-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty