Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-4-nitroaniline 95%

Supplier: Thermo Scientific Chemicals
Warning

H55174-14 H55174-22 H55174-06
AAH55174-14EA 141.99 USD InStock
AAH55174-14 AAH55174-06 AAH55174-22
2-Fluoro-4-nitroaniline 95%
2-Fluoro-4-nitroaniline


Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 122…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00034560
CAS Number: 369-35-7
EINECS: 206-719-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty