Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl (2-bromoethyl)carbamate 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(BOC-amino)ethyl bromide

H55211-03 H55211-06 H55211-MD
AAH55211-MDEA 21.02 USD InStock
AAH55211-MD AAH55211-06 AAH55211-03
tert-Butyl (2-bromoethyl)carbamate 96%
tert-Butyl (2-bromoethyl)carbamate


Formula: C₇H₁₄BrNO₂
MW: 224.10
Melting Pt: 30…32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683428
CAS Number: 39684-80-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty