Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-4-bromoaniline 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Bromoaniline, N-BOC protectedtert-Butyl (4-bromophenyl)carbamate

H55226-06 H55226-14 H55226-03
AAH55226-14EA 275.99 USD InStock
AAH55226-14 AAH55226-03 AAH55226-06
N-Boc-4-bromoaniline 97%
N-Boc-4-bromoaniline


Formula: C₁₁H₁₄BrNO₂
MW: 272.14 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD01006612
CAS Number: 131818-17-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty