Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxy-N,N-dimethylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

H55260-03 H55260-14 H55260-06
AAH55260-03EA 60.89 USD InStock
AAH55260-03 AAH55260-14 AAH55260-06
3-Methoxy-N,N-dimethylbenzylamine 98%
3-Methoxy-N,N-dimethylbenzylamine


Formula: C₁₀H₁₅NO
Boiling Pt: 105 °C/13 mm
Density: 0.984
Flash Pt: 96 °C(205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011530
CAS Number: 15184-99-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty