Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Boc-1-(5-bromo-2-pyridyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tert-Butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate5-Bromo-2-(4-tert-butoxycarbonylpiperazin-1-yl)pyridine

H55311-14 H55311-06 H55311-03
AAH55311-06EA 149.99 USD InStock
AAH55311-06 AAH55311-14 AAH55311-03
4-Boc-1-(5-bromo-2-pyridyl)piperazine 97%
4-Boc-1-(5-bromo-2-pyridyl)piperazine


Formula: C₁₄H₂₀BrN₃O₂
MW: 342.237 g/mol
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD07369772
CAS Number: 153747-97-8
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty