Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-7-bromofluorene 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 7-Bromo-9H-fluoren-2-amine

H55370-06 H55370-MD H55370-03
AAH55370-MDEA 26.09 USD InStock
AAH55370-MD AAH55370-06 AAH55370-03
2-Amino-7-bromofluorene 90%, Technical Grade
2-Amino-7-bromofluorene


Formula: C₁₃H₁₀BrN
MW: 260.134 g/mol
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00001127
CAS Number: 6638-60-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty