Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(3-Chlorobenzyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

H55409-06 H55409-14 H55409-03
AAH55409-14EA 199.8 USD InStock
AAH55409-14 AAH55409-03 AAH55409-06
1-(3-Chlorobenzyl)piperazine 98%
1-(3-Chlorobenzyl)piperazine


Formula: C₁₁H₁₅ClN₂
Boiling Pt: 120…123 °C/1.5 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02048053
CAS Number: 23145-91-7
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty