Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azidotri-n-butyltin(IV) 95%

Supplier: Thermo Scientific Chemicals
Danger

H55634-03 H55634-03 H55634-14 H55634-14 H55634-06 H55634-06
AAH55634-03EA 29.89 USD InStock
CAAAH55634-03 CAAAH55634-14 CAAAH55634-06 AAH55634-03 AAH55634-14 AAH55634-06
Azidotri-n-butyltin(IV) 95%
Azidotri-n-butyltin(IV)


Formula: C₁₂H₂₇N₃Sn
MW: 332.079 g/mol
Density: 1.212
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00216557
CAS Number: 17846-68-3
UN: 2788
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty