Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrofluorene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Nitro-9H-fluorene

H55744-14 H55744-14 H55744-06 H55744-06
AAH55744-06EA 41.29 USD InStock
CAAAH55744-06 CAAAH55744-14 AAH55744-06 AAH55744-14
2-Nitrofluorene 98%
2-Nitrofluorene


Formula: C₁₃H₉NO₂
MW: 211.22 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00001117
CAS Number: 607-57-8
EINECS: 210-138-5
ADR: 9,III
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty