Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Aminophenanthrene 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Phenanthren-9-amine

H55773-03 H55773-MD
AAH55773-MDEA 230.99 USD InStock
AAH55773-MD AAH55773-03
9-Aminophenanthrene 96%
9-Aminophenanthrene


Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00001177
CAS Number: 947-73-9
EINECS: 213-431-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty