Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromoanthranilic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-4-bromobenzoic acid

H55829-MD H55829-03 H55829-06
AAH55829-03EA 79.79 USD InStock
AAH55829-03 AAH55829-MD AAH55829-06
4-Bromoanthranilic acid 97%
4-Bromoanthranilic acid


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 230…234 °C
Storage Temperature: Ambient
MDL Number: MFCD03618454
CAS Number: 20776-50-5
EINECS: 244-025-7
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty