Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Benzoylpiperazine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Phenyl(piperazin-1-yl)methanone

H55896-06 H55896-03 H55896-14
AAH55896-03EA 33.69 USD InStock
AAH55896-03 AAH55896-14 AAH55896-06
1-Benzoylpiperazine 97%
1-Benzoylpiperazine


Formula: C₁₁H₁₄N₂O
MW: 190.24 g/mol
Boiling Pt: 145 °C/0.05 mm
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00810192
CAS Number: 13754-38-6
UN: 3263
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty