Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Tributylstannyl)thiophene 97%

Supplier: Thermo Scientific Chemicals
Danger

H55904-AE H55904-AC H55904-AA H55904-AA H55904-AC H55904-AE
AAH55904-AEEA 509.99 USD InStock
AAH55904-AE AAH55904-AC AAH55904-AA CAAAH55904-AC CAAAH55904-AE CAAAH55904-AA
2-(Tributylstannyl)thiophene 97%
2-(Tributylstannyl)thiophene


Formula: C₁₆H₃₀SSn
MW: 373.16
Boiling Pt: 155 °C/0.1 mm
Density: 1.175
Storage Temperature: Ambient
MDL Number: MFCD00192513
CAS Number: 54663-78-4
UN: 2788
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty