Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromothiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H55956-14 H55956-03 H55956-06
AAH55956-06EA 76.69 USD InStock
AAH55956-06 AAH55956-14 AAH55956-03
4-Bromothiobenzamide 97%
4-Bromothiobenzamide


Formula: C₇H₆BrNS
MW: 216.1 g/mol
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD04114360
CAS Number: 26197-93-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty