Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-4-(2-hydroxyethyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate4-(2-Hydroxyethyl)piperazine, N1-BOC protected

H55991-14 H55991-06 H55991-03
AAH55991-06EA 89.59 USD InStock
AAH55991-06 AAH55991-14 AAH55991-03
1-Boc-4-(2-hydroxyethyl)piperazine 97%
1-Boc-4-(2-hydroxyethyl)piperazine


Formula: C₁₁H₂₂N₂O₃
MW: 230.308 g/mol
Melting Pt: 38…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00728947
CAS Number: 77279-24-4
UN: 3259
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty