Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodine trichloride (Wijs'reagent) 97%

Supplier: Thermo Scientific Chemicals
Danger

H56005-06 H56005-22 H56005-14 H56005-22 H56005-14 H56005-06
AAH56005-22EA 354.99 USD InStock
CAAAH56005-22 CAAAH56005-14 CAAAH56005-06 AAH56005-22 AAH56005-06 AAH56005-14
Iodine trichloride (Wijs'reagent) 97%
Iodine trichloride (Wijs'reagent)


Formula: ICl₃
MW: 233.26 g/mol
Melting Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036291
CAS Number: 865-44-1
EINECS: 212-739-8
UN: 3260
ADR: "8",III
Merck Index: 13,05042
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty