Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Boc-3-methylamino azetidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

H56060-03 H56060-MD
AAH56060-03EA 467.99 USD InStock
AAH56060-03 AAH56060-MD
3-Boc-3-methylamino azetidine hydrochloride 95%
3-Boc-3-methylamino azetidine hydrochloride


Formula: C₉H₁₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12963843
CAS Number: 577777-20-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty