Synonyms:
2-Hydrazono-3-methylbenzothiazoline hydrochloride
, 2-Hydrazono-3-methyl-2,3-dihydrobenzo[d]thiazole hydrochloride hydrate
H56061-18
H56061-06
H56061-06
H56061-18
AAH56061-06EA
76.54
USD
InStock
CAAAH56061-06
CAAAH56061-18
AAH56061-06
AAH56061-18
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥98% (by HPLC)
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.
Caution: Incompatible with oxidizing agents. Store at room temperature.