Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Cysteine 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-Cysteine H-DL-Cys-OH

H56126-06 H56126-22 H56126-14 H56126-14 H56126-06 H56126-22
AAH56126-14EA 147.08 USD InStock
CAAAH56126-22 CAAAH56126-14 AAH56126-14 CAAAH56126-06 AAH56126-06 AAH56126-22
DL-Cysteine 96%
DL-Cysteine


Formula: C₃H₇NO₂S
MW: 121.16 g/mol
Melting Pt: 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004881
CAS Number: 3374-22-9
EINECS: 222-160-2
Merck Index: 12,02850
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty