Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-3-Hydroxytetrahydrofuran 98%

Supplier: Thermo Scientific Chemicals
Warning

H56476-14 H56476-14 H56476-06 H56476-03 H56476-03 H56476-06
AAH56476-14EA 298.99 USD InStock
AAH56476-14 AAH56476-03 AAH56476-06 CAAAH56476-06 CAAAH56476-14 CAAAH56476-03
(S)-(+)-3-Hydroxytetrahydrofuran 98%
(S)-(+)-3-Hydroxytetrahydrofuran


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 80 °C/15 mm
Density: 1.103
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064327
CAS Number: 86087-23-2
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty