Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Ethylenedioxythiophene 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,3-Dihydrothieno[3,4-b][1,4]dioxineEDOT

H56533-06 H56533-14 H56533-06 H56533-14
AAH56533-14EA 173.94 USD InStock
CAAAH56533-14 AAH56533-14 AAH56533-06 CAAAH56533-06
3,4-Ethylenedioxythiophene 97%
3,4-Ethylenedioxythiophene
Often used as a monomer to synthesize the conductive polymer PEDT


Formula: C₆H₆O₂S
MW: 142.17 g/mol
Boiling Pt: 193 °C
Density: 1.331
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD02093622
CAS Number: 126213-50-1
EINECS: 603-128-0
UN: 2810
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty