Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl cyclopropane-1,1-dicarboxylate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Diethyl 1,1-cyclopropanedicarboxylateDiethyl 1,1-cyclopropane dicarboxyliate

H56568-14 H56568-03 H56568-14 H56568-06 H56568-06 H56568-03
AAH56568-14EA 361.99 USD InStock
CAAAH56568-06 CAAAH56568-14 CAAAH56568-03 AAH56568-14 AAH56568-03 AAH56568-06
Diethyl cyclopropane-1,1-dicarboxylate
Diethyl cyclopropane-1,1-dicarboxylate


Formula: C₉H₁₄O₄
Boiling Pt: 94…96 °C/10 mm
Density: 1.055
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001283
CAS Number: 1559-02-0
EINECS: 216-321-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty