Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Usnic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (9bR)-2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione

H56612-14 H56612-06 H56612-06 H56612-14
AAH56612-06EA 59.09 USD InStock
AAH56612-06 AAH56612-14 CAAAH56612-06 CAAAH56612-14
(+)-Usnic acid 98%
(+)-Usnic acid


Formula: C₁₈H₁₆O₇
MW: 344.32 g/mol
Melting Pt: 201…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00016878
CAS Number: 7562-61-0
EINECS: 231-456-0
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty