Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Pinitol 95%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-O-Methyl-D-chiro-inositol

H56648-03 H56648-MD H56648-MD H56648-06 H56648-06 H56648-03
AAH56648-MDEA 38.49 USD InStock
AAH56648-MD CAAAH56648-MD CAAAH56648-06 AAH56648-06 CAAAH56648-03 AAH56648-03
D-Pinitol 95%
D-Pinitol


Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Melting Pt: 178…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00216659
CAS Number: 10284-63-6
Merck Index: 14,10361
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty