Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid sesquihydrate

H56704-03 H56704-03 H56704-06 H56704-06
AAH56704-03EA 19.29 USD InStock
CAAAH56704-06 AAH56704-03 CAAAH56704-03 AAH56704-06
(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 99%
(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate


Formula: C₁₀H₁₃NO₄·1.5H₂O
MW: 238.24 g/mol (211.22 anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149280
CAS Number: 41372-08-1
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty