Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromomethyl acetate 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid bromomethyl ester

H56755-06 H56755-09 H56755-06
AAH56755-06EA 256.99 USD InStock
AAH56755-06 AAH56755-09 CAAAH56755-06
Bromomethyl acetate 95%
Bromomethyl acetate


Formula: H₃CCOOCH₂Br
MW: 152.98 g/mol
Boiling Pt: 130…133 °C
Density: 1.56
Flash Pt: 57 °C(135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000170
CAS Number: 590-97-6
EINECS: 209-696-2
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty