Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Boc-1-oxa-6-azaspiro[3.3]heptane 96%

Supplier: Thermo Scientific Chemicals
Warning

H57040-ME H57040-MD
AAH57040-MEEA 401.82 USD InStock
AAH57040-ME AAH57040-MD
6-Boc-1-oxa-6-azaspiro[3.3]heptane 96%
6-Boc-1-oxa-6-azaspiro[3.3]heptane


Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Storage Temperature: Ambient
CAS Number: 1223573-41-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty