Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-n-Propylheptanoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H57414-03 H57414-MD
AAH57414-03EA 133.99 USD InStock
AAH57414-03 AAH57414-MD
2-n-Propylheptanoic acid 97%
2-n-Propylheptanoic acid


Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01723758
CAS Number: 31080-39-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty