Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-3-(bromomethyl)azetidine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl 3-(bromomethyl)azetidine-1-carboxylate

H57492-ME H57492-ME H57492-MD H57492-MD
AAH57492-MEEA 201.99 USD InStock
AAH57492-ME CAAAH57492-ME AAH57492-MD CAAAH57492-MD
1-Boc-3-(bromomethyl)azetidine 95%
1-Boc-3-(bromomethyl)azetidine


Formula: C₉H₁₆BrNO₂
MW: 250.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16556174
CAS Number: 253176-93-1
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty