Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-3-methylazetidine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl 3-ethynylazetidine-1-carboxylate

H57496-ME H57496-ME H57496-MD H57496-MD
AAH57496-MEEA 335.99 USD InStock
CAAAH57496-MD CAAAH57496-ME AAH57496-ME AAH57496-MD
1-Boc-3-methylazetidine 95%
1-Boc-3-methylazetidine


Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09951813
CAS Number: 1314984-00-3
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty