Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-3-(methylamino)azetidine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-butyl 3-(methylamino)azetidine-1-carboxylate

H57951-MD H57951-03
AAH57951-03EA 274.99 USD InStock
AAH57951-03 AAH57951-MD
1-Boc-3-(methylamino)azetidine 95%
1-Boc-3-(methylamino)azetidine


Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752496
CAS Number: 454703-20-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty