Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopropylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: 2-Propylzinc bromide

Packaged under Argon in resealable ChemSeal™ bottles.

ChemSeal
H58536-AD H58536-AD
AAH58536-ADEA 237.99 USD InStock
CAAAH58536-AD AAH58536-AD
Isopropylzinc bromide 0.5 M in THF
Isopropylzinc bromide
Isopropylzinc bromide is employed in croos-coupling reactions.

Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and water.


Formula: C₃H₇BrZn
MW: 188.373 g/mol
Density: 0.963
Storage Temperature: Ambient
MDL Number: MFCD06200639
CAS Number: 77047-87-1
UN: 3399
ADR: "4.3",III
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty