Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Adamantylzinc bromide solution 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger

Packaged under Argon in resealable ChemSeal™ bottles.

ChemSeal
H58595-AD
AAH58595-ADEA 269.99 USD InStock
AAH58595-AD
2-Adamantylzinc bromide solution 0.5 M in THF
2-Adamantylzinc bromide solution


Formula: C₁₀H₁₅BrZn
Density: 1.018
Storage Temperature: Ambient
MDL Number: MFCD01311401
CAS Number: 171860-65-4
UN: 3399
ADR: "4.3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty