Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Cyclopropyl-4-nitrobenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H58673-ME H58673-MC
AAH58673-MEEA 304.99 USD InStock
AAH58673-ME AAH58673-MC
N-Cyclopropyl-4-nitrobenzenesulfonamide 97%
N-Cyclopropyl-4-nitrobenzenesulphonamide


Formula: C₉H₁₀N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00458256
CAS Number: 549476-61-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty