Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Adamantanethiol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Mercaptoadamantane

H58958-MD H58958-03
AAH58958-03EA 113.87 USD InStock
AAH58958-03 AAH58958-MD
1-Adamantanethiol 98%
1-Adamantanethiol


Formula: C₁₀H₁₆S
MW: 168.298 g/mol
Melting Pt: 99…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00213530
CAS Number: 34301-54-7
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty