Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-Ser(tBu)-OH · DCHA

H59163-06 H59163-14 H59163-06 H59163-14
AAH59163-06EA 175.99 USD InStock
AAH59163-06 AAH59163-14 CAAAH59163-06 CAAAH59163-14
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt 98%
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt


Formula: C₂₄H₄₆N₂O₅
MW: 442.641 g/mol
Melting Pt: 161…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00065590
CAS Number: 18942-50-2
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty