Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Glutamic acid diethyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Glu(OEt)-OEt.HClDiethyl L-glutamateH-Glu(OEt)-OEt·HCl

H59551-06 H59551-14 H59551-06 H59551-14
AAH59551-14EA 122.99 USD InStock
CAAAH59551-06 AAH59551-14 AAH59551-06 CAAAH59551-14
L-Glutamic acid diethyl ester hydrochloride 98%
L-Glutamic acid diethyl ester hydrochloride


Formula: C₉H₁₈ClNO₄
MW: 239.696 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00012509
CAS Number: 1118-89-4
EINECS: 214-270-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty