Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl azetidin-3-ylcarbamate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Boc-amino)azetidine

H59556-MD H59556-MD H59556-ME H59556-ME
AAH59556-MDEA 84.49 USD InStock
AAH59556-MD AAH59556-ME CAAAH59556-MD CAAAH59556-ME
tert-Butyl azetidin-3-ylcarbamate 98%
tert-Butyl azetidin-3-ylcarbamate


Formula: C8H16N2O2
MW: 172.23 g/mol
Boiling Pt: 268 °C
Density: 1.06
Flash Pt: 116 °C(241 °F)
Storage Temperature: Ambient
MDL Number: MFCD01861759
CAS Number: 91188-13-5
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty