Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Fmoc-D-valine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-D-Val-OHD-Valine, N-FMOC protected

H59570-06 H59570-06 H59570-03 H59570-03
AAH59570-06EA 106.03 USD InStock
AAH59570-06 CAAAH59570-06 AAH59570-03 CAAAH59570-03
N-Fmoc-D-valine 98%
N-Fmoc-D-valine


Formula: C₂₀H₂₁NO₄
MW: 339.39 g/mol
Melting Pt: 143…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00062953
CAS Number: 84624-17-9
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty