Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω-Bromo-2,4-dichloroacetophenone 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Bromo-1-(2,4-dichlorophenyl)ethan-1-one2,4-Dichlorophenacyl bromideα-Bromo-2,4-dichloroacetophenone2-Bromo-2',4'-dichloroacetophenone2-Bromo-1-(2,4-dichlorophenyl)ethanone

H59615-06 H59615-06 H59615-03 H59615-03
AAH59615-03EA 78.69 USD InStock
CAAAH59615-06 CAAAH59615-03 AAH59615-03 AAH59615-06
ω-Bromo-2,4-dichloroacetophenone 95%
ω-Bromo-2,4-dichloroacetophenone


Formula: C₈H₅BrCl₂O
MW: 267.94 g/mol
Melting Pt: 25…29 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053005
CAS Number: 2631-72-3
EINECS: 220-116-7
UN: 1759
ADR: "8",III
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty