Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Boc-aminomethyl)azetidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-butyl N-(azetidin-3-ylmethyl)carbamate

H59816-MD H59816-ME H59816-MD H59816-ME
AAH59816-MDEA 142.99 USD InStock
AAH59816-MD AAH59816-ME CAAAH59816-MD CAAAH59816-ME
3-(Boc-aminomethyl)azetidine hydrochloride 95%
3-(Boc-aminomethyl)azetidine hydrochloride


Formula: C₉H₁₈N₂O₂
MW: 186.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861760
CAS Number: 91188-15-7
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty