Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Butyl-3-methyl-1H-imidazol-3-ium dibutyl phosphate

H59893-06 H59893-06 H59893-18 H59893-18
AAH59893-18EA 451.99 USD InStock
CAAAH59893-18 CAAAH59893-06 AAH59893-18 AAH59893-06
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 96%
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate


Formula: C₁₆H₃₃N₂O₄P
MW: 348.424 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265043
CAS Number: 663199-28-8
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty