Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Amino-7-bromo-9-methyl-7-deazapurine

H59897-03 H59897-MD H59897-MD H59897-03
AAH59897-03EA 370.99 USD InStock
CAAAH59897-03 AAH59897-03 CAAAH59897-MD AAH59897-MD
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 97%
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine


Formula: C₇H₇BrN₄
MW: 227.065 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486185
CAS Number: 1337532-51-0
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty