Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-tert-Butyl-2-aminopropanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-Ala-OtBu·HCltert-Butyl (R)-2-Aminopropionate Hydrochloride(R)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride

H59898-03 H59898-06 H59898-06 H59898-03
AAH59898-03EA 95.29 USD InStock
CAAAH59898-06 AAH59898-03 AAH59898-06 CAAAH59898-03
(R)-tert-Butyl-2-aminopropanoate hydrochloride 98%
(R)-tert-Butyl-2-aminopropanoate hydrochloride


Formula: C7H16ClNO2
MW: 181.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077087
CAS Number: 59531-86-1
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty