Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Methylpyrazole-5-carboxylic acid

H59899-06 H59899-06 H59899-03 H59899-03
AAH59899-03EA 113.99 USD InStock
AAH59899-03 AAH59899-06 CAAAH59899-06 CAAAH59899-03
3-Methyl-1H-pyrazole-5-carboxylic acid 97%
3-Methyl-1H-pyrazole-5-carboxylic acid


Formula: C5H6N2O2
MW: 126.11 g/mol
Melting Pt: 236…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00462235
CAS Number: 696-22-0
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty