Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Carotene 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Provitamin A2,2'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(1,3,3-trimethylcyclohex-1-ene)

H60106-06 H60106-03
AAH60106-06EA 125.78 USD InStock
AAH60106-06 AAH60106-03
β-Carotene 99%
beta-Carotene


Formula: C₄₀H₅₆
MW: 536.89 g/mol
Melting Pt: 178…179 °C
Density: 1.000
Flash Pt: 103 °C(217 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001556
CAS Number: 7235-40-7
EINECS: 230-636-6
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty