Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Asiatic acid 97%

Supplier: Thermo Scientific Chemicals

H60149-ME H60149-ME
AAH60149-MEEA 276.99 USD InStock
AAH60149-ME CAAAH60149-ME
Asiatic acid 97%
Asiatic acid


Formula: C₃₀H₄₈O₅
Melting Pt: 325…330 °C
Storage Temperature: Ambient
MDL Number: MFCD00238541
CAS Number: 464-92-6
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty