Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal 97%

Supplier: Thermo Scientific Chemicals

H60275-03 H60275-06 H60275-03 H60275-06
AAH60275-03EA 37.79 USD InStock
AAH60275-03 AAH60275-06 CAAAH60275-03 CAAAH60275-06
3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal 97%
3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal


Formula: C₂₄H₅₂O₄Si₃
Boiling Pt: 210 °C/1.2 mm
Density: 0.928
Flash Pt: >113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00171721
CAS Number: 79999-47-6
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty