Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-[Boc(methyl)aminomethyl]azetidine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(N-BOC-methylaminomethyl)azetidine2-Methyl-2-propanyl (3-azetidinylmethyl)methylcarbamateAzetidin-3-ylmethyl-methyl-carbamic acid tert-butyl ester

H60310-MD H60310-03 H60310-MD H60310-03
AAH60310-03EA 643.99 USD InStock
CAAAH60310-MD AAH60310-03 CAAAH60310-03 AAH60310-MD
3-[Boc(methyl)aminomethyl]azetidine 95%
3-[Boc(methyl)aminomethyl]azetidine


Formula: C₁₀H₂₀N₂O₂
MW: 200.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061967
CAS Number: 1053655-53-0
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty